EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Synthesis of curcumin analogues as potential antioxidant, cancer chemopreventive agents.
PMID 14760627
Abstract
New series of 3, 5-bis(substituted benzylidene)-4-piperidones, 2, 7-bis(substituted benzylidene)cycloheptanones, 1, 5-bis(substituted phenyl)-1, 4-pentadien-3-ones, 1, 7-bis(substituted phenyl)-1, 6-heptadien-3, 5-diones, 1, 1-bis(substituted cinnamoyl)-cyclopentanes, and 1, 1-bis(substituted cinnamoyl)cyclohexanes have been synthesized and tested for their antioxidant activity. Among the tested compounds, compounds II(4), II(9) II(10), II(11), V(1), and V(4) exhibited higher free radical scavenger activity with % inhibition values of 90.71, 91.24, 96.91, 94.26, 99.23, and 99.85%, respectively. Moreover, compound V(1) is the safest member toward peripheral multinuclear neutrophils (PMNs) with a % viability value of 91%. Detailed synthesis, spectroscopic, and biological data are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C99000
Cycloheptanone, 99%
C7H12O