EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Advances in the study of amygdalin].
PMID 14760843
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6005
Amygdalin, from apricot kernels, ≥99%
C20H27NO11
10050
Amygdalin, BioXtra, ≥97.0% (HPLC)
C20H27NO11