EMAIL THIS PAGE TO A FRIEND
Journal of cell science
The surface of articular cartilage contains a progenitor cell population.
PMID 14762107
Abstract
It is becoming increasingly apparent that articular cartilage growth is achieved by apposition from the articular surface. For such a mechanism to occur, a population of stem/progenitor cells must reside within the articular cartilage to provide transit amplifying progeny for growth. Here, we report on the isolation of an articular cartilage progenitor cell from the surface zone of articular cartilage using differential adhesion to fibronectin. This population of cells exhibits high affinity for fibronectin, possesses a high colony-forming efficiency and expresses the cell fate selector gene Notch 1. Inhibition of Notch signalling abolishes colony forming ability whilst activated Notch rescues this inhibition. The progenitor population also exhibits phenotypic plasticity in its differentiation pathway in an embryonic chick tracking system, such that chondroprogenitors can engraft into a variety of connective tissue types including bone, tendon and perimysium. The identification of a chondrocyte subpopulation with progenitor-like characteristics will allow for advances in our understanding of both cartilage growth and maintenance as well as provide novel solutions to articular cartilage repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
CP5, 08052101