EMAIL THIS PAGE TO A FRIEND
Bollettino chimico farmaceutico
[Pharmacology of escin, a saponin of Aesculus hippocastanum L. II. Pharmacodynamics of escin. Chapter II].
PMID 1476699
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1378 Escin, ≥95%, powder