Metabolism: clinical and experimental
Effect of R(+)alpha-lipoic acid on pyruvate metabolism and fatty acid oxidation in rat hepatocytes.
PMID 14767867
R-(+)-alpha-lipoic acid (R-LA) is the naturally occurring enantiomer of LA. It is a strong antioxidant and cofactor of key metabolic enzyme complexes catalyzing the decarboxylation of alpha-keto acids. Racemic LA (rac-LA) has shown promise in treating diabetic polyneuropathy, and some studies suggest that it improves glucose homeostasis in patients with type 2 diabetes. We examined the effects of R-LA on pyruvate metabolism and free fatty acid (FFA) oxidation in primary cultured hepatocytes isolated from 24-hour fasted rats. After overnight culture in serum-free medium, cells were pre-exposed to R-LA for 3 hours before assays. R-LA (25 to 200 micromol/L) significantly increased pyruvate oxidation ( approximately 2-fold at the highest dose tested) measured as (14)CO(2) production from [1-(14)C]pyruvate by the cells over 1 hour post-treatment. These effects correlated with proportional, significant increases in the activation state of the pyruvate dehydrogenase (PDH) complex. R-LA treatment inhibited glucose production from pyruvate by approximately 50% at 50 micromol/L R-LA and approximately 90% at 200 micromol/L. Palmitate oxidation was measured in hepatocytes cultured in the presence of albumin and physiological (0.1 mmol/L) or high (1.5 mmol/L) concentrations of FFA. The latter markedly enhanced FFA oxidation. R-LA treatment significantly inhibited FFA oxidation in both media, but was more effective in high FFA, where it reduced FFA oxidation by 48% to 82% at 25 to 200 micromol/L, respectively. Identical doses of R-LA did not affect FFA oxidation by L6 myotubes (a cell culture model for skeletal muscle) in either high or low FFA medium, but enhanced pyruvate oxidation. In conclusion, 3-hour exposure of primary cultured rat hepatocytes to R-LA at therapeutically relevant concentrations increased pyruvate oxidation, apparently by activation of the PDH complex, and decreased gluconeogenesis and FFA oxidation. These features may prove useful in the control of type 2 diabetes.
Related Materials
Product #
Molecular Formula
Add to Cart
(R)-(+)-α-Lipoic acid, ≥98.0% (HPLC)