EMAIL THIS PAGE TO A FRIEND
Annales pharmaceutiques francaises
[Experimental studies on conessine. VI. Comparison of experimental and clinical results of emetine and conessine].
PMID 14771682
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0095
Conessine, ≥97% (HPLC)
C24H40N2