EMAIL THIS PAGE TO A FRIEND
Zhonghua shen jing jing shen ke za zhi = Chinese journal of neurology and psychiatry
[Serum concentrations of amitriptyline and its metabolites and clinical effect in depressive patients].
PMID 1478132
Abstract
The serum concentrations of amitriptyline and its 3 metabolites--nortriptyline, 10-hydroxy-amitriptyline and 10-hydroxy-nortriptyline were determined in 18 depressive patients treated with amitriptyline for 6 weeks. The relationships between the serum concentrations and the clinical effects were statistically analyzed and showed significant correlations. The authors suggest that the total TAD serum concentration (280 ng/ml) can be used as the index for clinical efficacy, so far as the above data are concerned.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A115
Amitriptyline metabolite, (±)-E-10-hydroxylated-, analytical standard
C20H23NO