EMAIL THIS PAGE TO A FRIEND
Journal of general microbiology
A novel polysaccharide hydrolase cDNA (celD) from Neocallimastix patriciarum encoding three multi-functional catalytic domains with high endoglucanase, cellobiohydrolase and xylanase activities.
PMID 1479358
Abstract
A plant polysaccharide hydrolase cDNA, designated celD, was isolated from a cDNA library of the rumen fungus Neocallimastix patriciarum. The enzyme encoded by celD had endoglucanase, cellobiohydrolase and xylanase activities. Deletion analysis revealed that celD cDNA can be truncated to code for three catalytically active domains. Each domain had the same substrate specificity as the enzyme produced by the untruncated celD and also possessed cellulose-binding capacity. Substrate competition studies showed that carboxymethylcellulose and xylan appear to compete with methylumbelliferyl cellobioside for the same active site within each domain. Expression of celD transcript in the rumen fungus was constitutive and was not affected by the presence of cellulose in the culture medium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6018
4-Methylumbelliferyl β-D-cellobioside, glucanase substrate
C22H28O13