EMAIL THIS PAGE TO A FRIEND
Matrix (Stuttgart, Germany). Supplement
Inhibition of bacterial collagenase by phosphonamide peptides.
PMID 1480014
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5135
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
C23H32N4O7