EMAIL THIS PAGE TO A FRIEND
Soudni lekarstvi
[Tetramethyldiaminodiphenylmethane--a universal reagent for drug detection].
PMID 1480981
Abstract
Many a drug was identified by thin layer chromatography when using tetramethyldiaminodiphenylmethan (TDM) as a universal reagent subsequently to chlorination. This non-carcinogenic compound yields a characteristical tint with all the drugs and their metabolites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M44451
4,4′-Methylenebis(N,N-dimethylaniline), 98%
C17H22N2