EMAIL THIS PAGE TO A FRIEND
The American journal of physiology
Effect of neurohypophysectomy on antidiuretic activity of substituted 3-hydroxy cinchoninic acids in the rat.
PMID 14810900
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
174823
4-Quinolinecarboxylic acid, 97%
C10H7NO2