EMAIL THIS PAGE TO A FRIEND
Journal of the American Pharmaceutical Association. American Pharmaceutical Association
Arabic acid and its salts. I. The preparation of arabic acid and sodium arabate powder.
PMID 14813975
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3006 Arabic acid