EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Glycolaldehyde and glycolaldehyde phosphate as reaction components in enzymatic pentose formation.
PMID 14820350
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43517
Glycolaldehyde 2-phosphate calcium salt, ≥95.0% (TLC)
C2H3CaO5P