EMAIL THIS PAGE TO A FRIEND
British medical journal
Trial of a cinchoninic acid derivative in some collagen diseases.
PMID 14821421
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
174823
4-Quinolinecarboxylic acid, 97%
C10H7NO2