EMAIL THIS PAGE TO A FRIEND
Laval medical
[Experimentation with succinic dinitrile in psychiatry].
PMID 14825956
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160962
Succinonitrile, 99%
C4H4N2
14190
Succinonitrile, purum, ≥97.0% (GC)
C4H4N2