EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Haut- und Geschlechtskrankheiten
[Experiences with thianthrol salve in dermatology].
PMID 14836969
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122440
Thianthrene, 97%
C12H8S2