EMAIL THIS PAGE TO A FRIEND
Journal of dental research
A clinical study of the effect of tetradecylamine on plaque formation.
PMID 14841317
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T10200
Tetradecylamine, 95%
C14H31N