EMAIL THIS PAGE TO A FRIEND
Nordisk medicin
[Hypothalamic lesion with diabetes insipidus treated with cinchoninic acid].
PMID 14843451
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
174823
4-Quinolinecarboxylic acid, 97%
C10H7NO2