EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Cell spreading on collagen that has been exposed to reactive aldehydes.
PMID 1487029
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W342904
trans,trans-2,4-Hexadienal, ≥95%
C6H8O
180343
trans,trans-2,4-Hexadienal, 95%
C6H8O