EMAIL THIS PAGE TO A FRIEND
Molecular diversity
Microwave-enhanced liquid-phase synthesis of thiohydantoins and thioxotetrahydropyrimidinones.
PMID 14870850
Abstract
An efficient, microwave-assisted method for the liquid-phase combinatorial synthesis of 3,5-disubstituted thiohydantoins and 3,5-disubstituted 2-thioxotetrahydropyrimidin-4-ones has been developed. In synthesizing thiohydantoins, Fmoc-protected amino acids were coupled with polymer support and then deprotected to give primary amines. While in synthesizing thioxotetrahydropyrimidinones, 3-chloropropionyl chloride was immobilized to the support and subsequently reacted with various amines to form secondary amines. The PEG bound primary/secondary amines then were incorporated with various isothiocyanates to give thiourea intermediates and concomitant cyclization/cleavage steps occurred under mild basic condition. The desired products were then liberated from the soluble matrix in good yield and purity. All reactions described here were performed under microwave irradiation.