EMAIL THIS PAGE TO A FRIEND
Chemical reviews
Synthetic analogues and reaction systems relevant to the molybdenum and tungsten oxotransferases.
PMID 14871153
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357189
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis
W
267546
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
W
357197
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
W
267538
Tungsten, foil, thickness 0.05 mm, ≥99.9% trace metals basis
W
267511
Tungsten, powder, 12 μm, 99.9% trace metals basis
W
357421
Tungsten, powder, 10 μm, ≥99.99% trace metals basis
W
510106
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
W
267562
Tungsten, wire, diam. 1.0 mm, 99.99% trace metals basis
W
267554
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
W
356972
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
W
767522
Tungsten, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
W
GF12269998
Tungsten, wire reel, 10m, diameter 0.08mm, clean, 99.95%
W
GF81410509
Tungsten, wire reel, 10m, diameter 0.08mm, gold plated, 99.95%
W
GF09514830
Tungsten, wire reel, 10m, diameter 0.125mm, clean, 99.95%
W
GF02629571
Tungsten, foil, 0.5m coil, thickness 0.035mm, coil width .5mm, as rolled, 99.95%
W
GF35593555
Tungsten, foil, 0.5m coil, thickness 0.05mm, coil width 2mm, as rolled, 99.95%
W
GF90049574
Tungsten, foil, 0.5m coil, thickness 0.2mm, coil width .6mm, as rolled, 99.95%
W
GF83538490
Tungsten, foil, 1m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
W
GF60432207
Tungsten, foil, 1m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
W
GF91197748
Tungsten, foil, 1m coil, thickness 0.1mm, coil width .6mm, as rolled, 99.95%
W
GF99741741
Tungsten, foil, 2m coil, thickness 0.025mm, coil width .7mm, as rolled, 99.95%
W
GF66018999
Tungsten, foil, 2m coil, thickness 0.04mm, coil width 0.7mm, as rolled, 99.95%
W
GF17648589
Tungsten, foil, 2m coil, thickness 0.1mm, coil width .6mm, as rolled, 99.95%
W
GF18728961
Tungsten, microfoil, disks, 25mm, thinness 0.1μm, specific density 182μg/cm2, permanent mylar 3.5μm support, 99.95%
W
GF22706210
Tungsten, rod, 1000mm, diameter 5.0mm, centerless ground, 99.95%
W
GF01295502
Tungsten, rod, 100mm, diameter 20.0mm, centerless ground, 99.95%
W
GF30443554
Tungsten, rod, 18mm, diameter 20.0mm, centerless ground, 99.95%
W
GF33990273
Tungsten, rod, 200mm, diameter 3.0mm, centerless ground, 99.95%
W
GF38415023
Tungsten, rod, 500mm, diameter 10.0mm, centerless ground, 99.95%
W
GF21910700
Tungsten, rod, 500mm, diameter 8.0mm, centerless ground, 99.95%
W