EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
DNA photosensitization by indoprofen - is DNA damage photoinduced by indoprofen or by its photoproducts?
PMID 14872241
Abstract
The in vitro photosensitizing activity of indoprofen, a non-steroidal anti-inflammatory drug, toward DNA has been studied by gel sequencing experiments using (32)P-end labelled synthetic oligonucleotides in phosphate buffered solution. Upon irradiation at [small lambda] > 320 nm, piperidine-sensitive lesions were induced in single- and double-stranded DNA, exclusively at the position of guanine bases. In single-stranded DNA, all G sites were modified. This pattern of photooxidative damage without isotopic effect in deuterium oxide, is characteristic of a Type I mechanism involving electron transfer from the base to the excited drug. In duplex DNA, a Type I process was also observed since selective DNA breakage occurred with high selectivity at 5[prime or minute]-G of a 5[prime or minute]-GG-3[prime or minute]sequence. When the oligonucleotide displays TT sites, an energy transfer process becomes predominant, giving rise to the formation of thymine dimers as evidenced by using T4 endonuclease V. Moreover, the methyl ester of indoprofen has been synthesized in order to study the influence of the indoprofen photochemical properties in DNA photosensitization. The poor efficiency of this compound shows that the drug itself is not directly implicated in DNA photodamage and seems to imply the involvement of indoprofen photoproducts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I3132
Indoprofen, analytical standard
C17H15NO3