EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Optimal aminoglycoside dosing regimen for penicillin-tobramycin synergism in experimental Streptococcus adjacens endocarditis.
PMID 1489184