EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
Lead tetraacetate-thiocarbohydrazide-silver proteinate method for light microscopy of polysaccharides.
PMID 1489842
Abstract
Lead tetraacetate-thiocarbohydrazide-silver proteinate reaction sequence for light microscopy of polysaccharides was evaluated on Carnoy's fixed rat liver sections. The results of this evaluation suggest that, on the light microscopic level, the lead tetraacetate-thiocarbohydrazide-silver proteinate method may serve as a practical and histochemically specific alternative to the lead tetraacetate-Schiff reaction for the localization of tissue carbohydrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185191
Lead(IV) acetate, reagent grade, 95%
C8H12O8Pb
223220
Thiocarbohydrazide, 98%
CH6N4S