EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology, therapy, and toxicology
Pharmacokinetics and efficacy of carbamoylphenoxyacetid acid.
PMID 1490812
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
167428
(2-Carbamoylphenoxy)acetic acid sodium salt, 97%
C9H8NNaO4