EMAIL THIS PAGE TO A FRIEND
Nature
Bacterial enzyme preparations oxidizing inositol and their inhibition by colchicine.
PMID 14910609
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80999
1L-epi-2-Inosose, ≥98.0% (HPLC)
C6H10O6