EMAIL THIS PAGE TO A FRIEND
Marseille medical
[Conessine in therapy of intestinal amebiasis].
PMID 14918670
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0095
Conessine, ≥97% (HPLC)
C24H40N2