EMAIL THIS PAGE TO A FRIEND
Minerva urologica e nefrologica = The Italian journal of urology and nephrology
[Comparison of efficacy and tolerability among 3 quinolones in the treatment of lower urinary tract infections (cinoxacin versus ciprofloxacin and pefloxacin].
PMID 1492275
Abstract
A clinical trial was carried out in 150 patients with lower urinary tract infections in order to compare the efficacy and tolerability of three quinolone drugs (cinoxacin, ciprofloxacin, pefloxacin). The satisfactory therapeutic efficacy of all three drugs is underlined, as is the excellent tolerability of cinoxacin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8645
Cinoxacin
C12H10N2O5