EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from Kathon 893.
PMID 1493706
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46078
2-Octyl-4-isothiazolin-3-one, PESTANAL®, analytical standard
C11H19NOS