EMAIL THIS PAGE TO A FRIEND
Comparative hepatology
Cytotoxic reactions of CC531s towards liver sinusoidal endothelial cells: a microscopical study.
PMID 14960201
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4292
3,3′-Dioctadecyloxacarbocyanine perchlorate
C53H85ClN2O6