EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The catalytic and kinetic mechanisms of NADPH-dependent alkenal/one oxidoreductase.
PMID 14966122
Abstract
NADPH-dependent alkenal/one oxidoreductase (AOR) from the rat is a phase 2/antioxidative enzyme that is known to catalyze the reduction of the carbon-carbon double bond of alpha,beta-unsaturated aldehydes and ketones. It is also known for its leukotriene B(4) 12-hydroxydehydrogenase activity. In order to begin to understand these dual catalytic activities and validate its classification as a reductase of the medium-chain dehydrogenase/reductase family, an investigation of the mechanism of its NADPH-dependent activity was undertaken. Recombinant AOR and a 3-nonen-2-one substrate were used to perform steady-state initial velocity, product inhibition, and dead end inhibition experiments, which elucidated an ordered Theorell-Chance kinetic mechanism with NADPH binding first and NADP(+) leaving last. A nearly 20-fold preference for NADPH over NADH was also observed. The dependence of kinetic parameters V and V/K on pH suggests the involvement of a general acid with a pK of 9.2. NADPH isomers stereospecifically labeled with deuterium at the 4-position were used to determine that AOR catalyzes the transfer of the pro-R hydride to the beta-carbon of an alpha,beta-unsaturated ketone, illudin M. Two-dimensional nuclear Overhauser effect NMR spectra demonstrate that this atom becomes the R-hydrogen at this position on the metabolite. Using [4R-(2)H]NADPH, small primary kinetic isotope effects of 1.16 and 1.73 for V and V/K, respectively, were observed and suggest that hydride transfer is not rate-limiting. Atomic absorption spectroscopy indicated an absence of Zn(2+) from active preparations of AOR. Thus, AOR fits predictions made for medium-chain reductases and bears similar characteristics to well known medium-chain alcohol dehydrogenases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262536
trans-3-Nonen-2-one, 95%
C9H16O