EMAIL THIS PAGE TO A FRIEND
Biophysical chemistry
EXAFS studies of structural changes in fragile glasses of zinc nitrate and nickel nitrate hydrates.
PMID 14967238
Abstract
The ionic structures of aqueous solutions of two sets of transition metal nitrates have been studied in the liquid and glass states by EXAFS spectroscopy. Experiments were carried out on Zn(NO3)2.xH2O, with x=2, 6, 12 and NiNO.9HO over the temperature range 30