EMAIL THIS PAGE TO A FRIEND
European radiology
Dose reduction in skeletal and chest radiography using a large-area flat-panel detector based on amorphous silicon and thallium-doped cesium iodide: technical background, basic image quality parameters, and review of the literature.
PMID 14968260
Abstract
The two most frequently performed diagnostic X-ray examinations are those of the extremities and of the chest. Thus, dose reduction in the field of conventional skeletal and chest radiography is an important issue and there is a need to reduce man-made ionizing radiation. The large-area flat-panel detector based on amorphous silicon and thallium-doped cesium iodide provides a significant reduction of radiation dose in skeletal and chest radiography compared with traditional imaging systems. This article describes the technical background and basic image quality parameters of this 43 x 43-cm digital system, and summarizes the available literature (years 2000-2003) concerning dose reduction in experimental and clinical studies. Due to its high detective quantum efficiency and dynamic range compared with traditional screen-film systems, a dose reduction of up to 50% is possible without loss of image quality.