EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Re: Selenium supplementation and secondary prevention of nonmelanoma skin cancer in a randomized trial.
PMID 14970283
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257710
Selenium sulfide
S2Se