EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Potent nonclassical nucleoside antiviral drugs based on the N,N-diarylformamidine concept.
PMID 14971898
Abstract
New formamidine-3TC (3TC = 2',3'-dideoxy-3'-thiacytidine) analogues have been synthesized through various methods, and their antiviral activities (HIV, HBV) have been evaluated in vitro. Anti-HIV-1 in acutely infected MT-4 cells and peripheral blood monocellular cells (PBMCs) showed that compounds substituted by N,N-diarylformamidine side chains at the 4-N nucleic base position (compounds 3 and 8-11) had at least equivalent anti-HIV activity as 3TC (EC50 = 0.5 and 11.6 microM, respectively). Moreover, the newly synthesized compounds demonstrated higher anti-HBV activity (EC50 ranging from 0.01 to 0.05 microM) compared to the parent nucleoside 3TC (EC50 = 0.2 microM). It should be underlined that these new promising derivatives inhibited HIV in cells of a macrophage lineage, which are known to be cellular reservoir for HIV. These results were particularly of interest, since the antiviral activities appeared not to be mediated through the formamidine bond hydrolysis and consequently the release of free 3TC. These new analogue series were found to be highly stable to hydrolysis even after prolonged incubation in different biological media (t(1/2) ranged from 48 to 120 h). This enzymatic stability, coupled to the fact that no delay in the antiviral response was observed compared to the free 3TC antiviral response, suggest that this new N,N-diarylformamidine nucleoside series should not be considered as classical prodrugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F15803
Formamidine acetate salt, 99%
CH4N2 · C2H4O2
268607
Formamidine hydrochloride, 97%
CH4N2 · HCl