EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Cutin and suberin monomers are membrane perturbants.
PMID 14972629
Abstract
The interaction between cutin and suberin monomers, i.e., omega -hydroxylpalmitic acid, alpha, omega -hexadecanedioic acid, alpha, omega --hexadecanediol, 12-hydroxylstearic acid, and phospholipid vesicles biomimicking the lipid structure of plant cell membranes has been studied by optical and transmission electron microscopy, quasielastic light scattering, differential scanning calorimetry, and (31)P solid-state NMR. Monomers were shown to penetrate model membranes until a molar ratio of 30%, modulating their gel to fluid-phase transition, after which monomer crystals also formed in solution. These monomers induced a decrease of the phospholipid vesicle size from several micrometers to about 300 nm. The biological implications of these findings are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
177490
16-Hydroxyhexadecanoic acid, 98%
C16H32O3