EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[What did the ALLHAT study bring? To do the one without foregoing the other].
PMID 14974327
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0915 Xipamide, ≥98% (HPLC)
C15H15ClN2O4S