EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Suppression of human pancreatic cancer cell proliferation by AGN194204, an RXR-selective retinoid.
PMID 14976133
Abstract
Retinoids may be useful agents for the treatment of pancreatic cancer. However, retinoic acid receptor (RAR)-selective retinoids produce unwanted side effects. In contrast, retinoid X receptor (RXR)-selective retinoids produce fewer side effects; however, it was not known whether RXR-selective retinoids could reduce pancreatic tumor cell proliferation. In the present study, the novel RXR-selective retinoid, AGN194204, was compared with that of other retinoids for the ability to suppress pancreatic cancer cell proliferation. We treated various pancreatic cancer cell lines with receptor-selective ligands and cytotoxic agents and monitored the effects on cell proliferation, markers of apoptosis and cell cycle. Our results indicate that AGN194204, at concentrations >10 nM, inhibits proliferation of MIA PaCa-2 and BxPC-3 cells but not the proliferation of AsPC-1 cells. Moreover, in BxPC-3 and MIA PaCa-2 cells, AGN194204 was 10-100 times more effective than RAR-selective retinoids. AGN194204-dependent suppression of MIA PaCa-2 cell proliferation is associated with reduced cyclin E and cyclin-dependent kinase 6 (cdk6) level, but cyclin D1, cdk2 and cdk4 content is not altered. In addition, p27 level increases 2-fold. The RXR-selective antagonist, AGN195393, reverses the AGN194204-dependent growth inhibition and the decline in cyclin E and cdk6 levels. In contrast, these changes are not reversed by treatment with the RAR antagonist, AGN193109. AGN194204 did not appear to alter cell apoptosis as measured by change in cleavage of procaspase-3, -8 or -9. We also examined the effects AGN194204 co-treatment with cytotoxic agents. Treatment of MIA PaCa-2 cells with AGN194204 + cisplatin, gemcitabine, 5-fluorouracil, interferon (IFN)alpha or IFNgamma resulted in an additive but not synergistic reduction in MIA PaCa-2 cell number. These results indicate that AGN194204, an RXR-selective retinoid, is a more effective inhibitor of pancreatic cell proliferation than the RAR-selective retinoids, and further indicate that AGN194204 produces an additive reduction in cell number when given with other agents. Our results suggest that RXR-selective ligands, which are less toxic than RAR-selective ligands, may be suitable agents for the treatment of pancreatic cancer.