EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Glyoxalase II of African trypanosomes is trypanothione-dependent.
PMID 14976196
Abstract
The glyoxalase system is a ubiquitous pathway catalyzing the glutathione-dependent detoxication of ketoaldehydes such as methylglyoxal, which is mainly formed as a by-product of glycolysis. The gene encoding a glyoxalase II has been cloned from Trypanosoma brucei, the causative agent of African sleeping sickness. The deduced protein sequence contains the highly conserved metal binding motif THXHXDH but lacks three basic residues shown to fix the glutathione-thioester substrate in the crystal structure of human glyoxalase II. Recombinant T. brucei glyoxalase II hydrolyzes lactoylglutathione, but does not show saturation kinetics up to 5 mm with the classical substrate of glyoxalases II. Instead, the parasite enzyme strongly prefers thioesters of trypanothione (bis(glutathionyl)spermidine), which were prepared from methylglyoxal and trypanothione and analyzed by high performance liquid chromatography and mass spectrometry. Mono-(lactoyl)trypanothione and bis-(lactoyl)trypanothione are hydrolyzed by T. brucei glyoxalase II with k(cat)/K(m) values of 5 x 10(5) m(-1) s(-1) and 7 x 10(5) m(-1) s(-1), respectively, yielding d-lactate and regenerating trypanothione. Glyoxalase II occurs in the mammalian bloodstream and insect procyclic form of T. brucei and is the first glyoxalase II of the order of Kinetoplastida characterized so far. Our results show that the glyoxalase system is another pathway in which the nearly ubiquitous glutathione is replaced by the unique trypanothione in trypanosomatids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10107085001 L-Lactate Dehydrogenase (L-LDH), from hog muscle