Molecular pharmacology
SB-505124 is a selective inhibitor of transforming growth factor-beta type I receptors ALK4, ALK5, and ALK7.
PMID 14978253
Clinically, there is a great need for small molecule inhibitors that could control pathogenic effects of transforming growth factor (TGF-beta) and/or modulate effects of TGF-beta in normal responses. Inhibition of TGF-beta signaling would be predicted to enhance re-epithelialization of cutaneous wounds and reduce scarring fibrosis. Selective small molecule inhibitors of the TGF-beta signaling pathway developed for therapeutics will also be powerful tools in experimentally dissecting this complex pathway, especially its cross-talk with other signaling pathways. In this study, we characterized 2-(5-benzo[1,3]dioxol-5-yl-2-tert-butyl-3H-imidazol-4-yl)-6-methylpyridine hydrochloride (SB-505124), a member of a new class of small molecule inhibitors related to imidazole inhibitors of p38, which inhibit the TGF-beta type I receptor serine/threonine kinase known as activin receptor-like kinase (ALK) 5. We demonstrate that this compound selectively and concentration-dependently inhibits ALK4-, ALK5-, and ALK 7-dependent activation of downstream cytoplasmic signal transducers, Smad2 and Smad3, and of TGF-beta-induced mitogen-activated protein kinase pathway components but does not alter ALK1, ALK2, ALK3 or ALK6-induced Smad signaling. SB-505124 also blocks more complex endpoints of TGF-beta action, as evidenced by its ability to abrogate cell death caused by TGF-beta1 treatment. SB-505124 is three to five times more potent than a related ALK5 inhibitor described previously, SB-431542.
Related Materials
Product #
Molecular Formula
Add to Cart
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
C20H21N3O2 · xHCl · yH2O