EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Recently synthesized class of vinylphosphonates as potent matrix metalloproteinase (MMP-2) inhibitors.
PMID 14981663
Abstract
An in vitro evaluation of MMP-2 inhibitors for a series of novel vinylphosphonic acids and phosphonic esters that contain various functional groups, shows that various types exhibit excellent efficiency, and points towards potent, promising compounds. Other types displayed relatively weak activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
396311
Vinylphosphonic acid, 97%
C2H5O3P
672068
Vinylphosphonic acid, ≥90.0% (T)
C2H5O3P