EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
Nifedipine loaded chitosan microspheres prepared by emulsification phase-separation.
PMID 14989642
Abstract
A high yield of nifedipine-chitosan microspheres could be obtained using an emulsification phase-separation method. A high level of entrapment of nifedipine in the microspheres was achieved. The microspheres exhibited excellent swelling properties. Differential scanning calorimetry, X-ray diffractometry, and scanning electron microscopy confirmed that at 1.84% loading, nifedipine was dispersed molecularly. The microspheres exhibited faster release at low loadings compared to high loadings. Fitting the data to the coupled Fickian/case II equation, showed that at low loadings polymer relaxation coefficients (k2) were high. As the polymer content increased in the microspheres, the value of n (diffusional exponent characteristic of the release mechanism) approached one, which is indicative of zero order.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7135
Span® 85