EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Severe cholestatic hepatitis induced by pyritinol.
PMID 15001508
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7171
Pyrithioxin, analytical standard
C16H20N2O4S2
P5156
Pyrithioxin dihydrochloride, analytical standard
C16H20N2O4S2 · 2HCl