EMAIL THIS PAGE TO A FRIEND
Biocell : official journal of the Sociedades Latinoamericanas de Microscopia Electronica ... et. al
Ultrastructural effects of two phthalocyanines in CHO-K1 and HeLa cells after laser irradiation.
PMID 15002747
Abstract
The effects of Photodynamic Therapy using 2nd generation photosensitizers have been widely investigated aiming clinical application treatment of solid neoplasms. In this work, ultrastructure changes caused by the action of two 2nd generation photosensitizers and laser irradiation on CHO-K1 and HeLa (neoplastic) cells were analyzed by transmission electron microscopy. Aluminum phthalocyanine chloride, aluminum phthalocyanine tetrasulfonate chloride and radiation from a semiconductor laser at a fluency of 0.5 J/cm2 (Power=26 mW; lambda=.670 nm) were used. The results showed induction of apoptosis. Such alterations where observed in HeLa but not in CHO-K1 cells after Aluminum phthalocyanine tetrasulfonate chloride (AlPcS4, photodynamic treatment. The Aluminum phthalocyanine chloride (AlPc) photodynamic treatment induced necrosis on the neoplastic cell line, and cytoplasm and nuclear alterations on the normal cell line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
362530
Aluminum phthalocyanine chloride, Dye content ~85 %
C32H16AlClN8