EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Crystal structure communications
(Z)-3-(1-Methyl-1H-indol-3-yl)-2-(thiophen-3-yl)acrylonitrile.
PMID 15004388
Abstract
The title compound, C16H12N2S, has been synthesized by base-catalyzed condensation of 1-methylindole-3-carboxaldehyde with thiophene-3-acetonitrile. The product assumes an approximately planar Z configuration. The molecule has a thienyl-ring flip disorder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357987
1-Methylindole-3-carboxaldehyde, 97%
C10H9NO