EMAIL THIS PAGE TO A FRIEND
Alternative medicine review : a journal of clinical therapeutic
Gamma-linolenic acid (GLA). Monograph.
PMID 15005646
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2378
γ-Linolenic acid, ≥99%, liquid
C18H30O2
62174
γ-Linolenic acid, analytical standard
C18H30O2