EMAIL THIS PAGE TO A FRIEND
The Journal of endocrinology
NO-1886 decreases ectopic lipid deposition and protects pancreatic beta cells in diet-induced diabetic swine.
PMID 15012594
Abstract
The synthetic compound NO-1886 (ibrolipim) is a lipoprotein lipase activator that has been proven to be highly effective in lowering plasma triglycerides. Recently, we found that NO-1886 also reduced plasma free fatty acids and glucose in high-fat/high-sucrose diet-induced diabetic rabbits. In the current study, we investigated the effects of NO-1886 treatment on ectopic lipid deposition and the islet pathology in miniature swine fed a high-fat/high-sucrose diet. Our results showed that feeding this diet to miniature swine caused insulin resistance, increased lipid deposition in non-adipose tissue, such as in the heart, skeletal muscle, liver and pancreas, and also caused pancreatic beta cell damage. However, supplementing 1% NO-1886 (200 mg/kg per day) into the high-fat/high-sucrose diet decreased ectopic lipid deposition, improved insulin resistance, and alleviated the beta cell damage. These results suggest that improvement of lipid disorder, non-adipose tissue steatosis and insulin resistance may be very important for the protection of beta cell damage. Therefore, NO-1886 is potentially beneficial for the treatment of insulin-resistance syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1540
NO-1886
C19H20BrN2O4P