EMAIL THIS PAGE TO A FRIEND
Anti-cancer drugs
Combretastatin A4 phosphate.
PMID 15014350
Abstract
Combretastatin A4 phosphate (CA4P) is a water-soluble prodrug of combretastatin A4 (CA4). The vascular targeting agent CA4 is a microtubule depolymerizing agent. The mechanism of action of the drug is thought to involve the binding of CA4 to tubulin leading to cytoskeletal and then morphological changes in endothelial cells. These changes increase vascular permeability and disrupt tumor blood flow. In experimental tumors, anti-vascular effects are seen within minutes of drug administration and rapidly lead to extensive ischemic necrosis in areas that are often resistant to conventional anti-cancer treatments. Following single-dose administration a viable tumor rim typically remains from which tumor regrowth occurs. When given in combination with therapies targeted at the proliferating viable rim, enhanced tumor responses are seen and in some cases cures. Results from the first clinical trials have shown that CA4P monotherapy is safe and reduces tumor blood flow. There has been some promising demonstration of efficacy. CA4P in combination with cisplatin is also safe. Functional imaging studies have been used to aid the selection of doses for phase II trials. Both dynamic contrast-enhanced magnetic resonance imaging (DCE-MRI) and positron emission tomography can measure the anti-vascular effects of CA4P in humans. This review describes the background to the development of CA4P, its proposed mechanism of action, the results from the first clinical trials with CA4P and the role of imaging techniques in its clinical development.