EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
NTP-CERHR Expert Panel Report on the reproductive and developmental toxicity of 2-bromopropane.
PMID 15019719
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B78114
2-Bromopropane, 99%
C3H7Br
59370
2-Bromopropane, purum, ≥99.0% (GC)
C3H7Br