EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Association of titin and myosin heavy chain in developing skeletal muscle.
PMID 1502160
Abstract
To understand molecular interactions that organize developing myofibrils, we examined the biosynthesis and interaction of titin and myosin heavy chain in cultures of developing muscle. Use of pulse-labeling, immunoprecipitation, and a reversible cross-linking procedure demonstrates that within minutes of synthesis, titin and myosin heavy chain can be chemically cross-linked into very large, detergent-resistant complexes retaining many features of intact myotubes. These complexes, predominantly of titin and myosin, occur very early in myofibrillogenesis as well as later. These data suggest that synthesis and assembly of titin and myosin are temporally and spatially coordinated in nascent myofibrils and support the hypothesis that titin molecules help to organize sarcomere formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3257
Ethylene glycol-bis(succinic acid N-hydroxysuccinimide ester), powder
C18H20N2O12